Visual Colour Kits, Free Chlorine
Supplier: Hach
1454201 1454202
104085-494EA 245.57 USD
104085-494 104085-496
Visual Colour Kits, Free Chlorine
Environmental Test Kits
Simple, economical, rapid visual color determination.
DPD visual test kit method for free chlorine, which can be used to accurately measure the free chlorine levels in the field or in the lab.
Delivery information: Kit includes one reagent bottle, stopper, color comparator box, color disc, tube, instruction sheet, and a carrying case.