Powder Pillows, Pathoscreen Medium Presence/Absence
Supplier: Hach
2610696
2610796
104086-478PK
145.38
USD
104086-478
104086-480
Powder Pillows, Pathoscreen Medium Presence/Absence
Media for Microbiology
Prepared Media for Microbiology
These are used to detect waterborne pathogens associated with fecal contamination of water sources.
- Simple presence-absence method eliminates need for an incubator in tropical and semi-tropical climates
- Complete Solutions for testing drinking water