Pocket Pro™ / Pocket Pro™+ Pocket pH and ORP Testers
Supplier: Hach
9531100 9532000 9532100 9531000
104087-098EA 540 USD
104087-098 104087-110 104087-114 104088-374
Pocket Pro™ / Pocket Pro™+ Pocket pH and ORP Testers
Electrochemistry Meters pH and Multi-Parameter Meters
These testers with integrated or replaceable sensor measure pH or ORP in a broad range of water applications. Ideal for field measurements in beverages, industrial water, wasterwater and swimming pool water.
  • pH testers with pH glass sensing element
  • Easy calibration steps and built-in diagnostics for pH testers
  • Easy-to-replace AAA batteries and a convenient sample cup make maintenance and measurements simple
  • Large LCD screen is easy to read
  • Intuitive user interface
  • Pocket Pro™+ models feature a replaceable sensor and backlit display
  • One year warranty on meter and sensor
Delivery information: pH tester supplied with 4 AAA batteries, lanyard, calibration solution and quick start guide. ORP tester is supplied with 4 AAA batteries, lanyard and quick start guide.