Ampoules m-Endo Broth
Supplier: Hach
2373520
2373550
104086-166PK
95.31
USD
104086-166
104086-168
Ampoules m-Endo Broth
Media for Microbiology
Prepared Media for Microbiology
m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and brilliant green bile broth tubes. Samples that test positive for total coliform bacteria should be further analyzed for the presence of faecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.
- Save valuable time and money using prepared media for membrane filtration methods