BART Tests for the Presence or Absence of Harmful Bacteria
Supplier: Hach
2619309 2619407 2432409 2432509 2432609 2478409 2490409 2490427
104086-488PK 389.87 USD
104086-488 104086-490 104086-244 104086-248 104086-252 104086-328 104086-338 104086-340
BART Tests for the Presence or Absence of Harmful Bacteria
Microbiology Test Systems Enzymatic Tests
Biological Activity Reaction Test (BART) biodetectors are microbiological diagnostic tools used to identify the presence of process disruptors. Tests for many types of bacteria are available, including sulfate-reducing bacteria, heterotropic acid, nitrifying bacteria, pool and spa bacteria, blue-green algae, and more.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment
Test results are distinct and easy to read: Observe the tubes every day to detect response indicators such as changes in color, turbidity, sediment, precipitation and colorful formations, such as disc and bubbles. Compare the observed with the comparison chart provided with the manual to identify the presence or absence of bacteria. Traditional methods can be used to confirm the identification of specific bacteria.
Note: The POOL test SLYM require a UV lamp.