TNTplus™ Sulphate Kit
Supplier: Hach
TNT864 TNT865
104087-858PK 188.52 USD
104087-858 104087-860
TNTplus™ Sulphate Kit
Environmental Test Kits
This kit is used for the the determination of sulphate by the turbidimetric method using TNTplus vials.
  • Reduce errors with bar coded vials and RFID
  • Innovative vials with DosiCaps
  • No reagent blank necessary