TNTplus™ Test Kits, Phosphorus
Supplier: Hach
TNT844 2742545 NT843
104087-838PK 150.62 USD
104087-838 104088-286 104088-292
TNTplus™ Test Kits, Phosphorus
Environmental Test Kits
TNT Plus kit is used for the efficient and accurate determination of reactive (ortho) and total phosphorus (phosphate) using TNTplus® vials.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary