Nitrite Kits, TNT and TNT plus Kit
Supplier: Hach
2608345 TNT839
104086-472EA 149.54 USD
104086-472 104088-668
Nitrite Kits, TNT and TNT plus Kit
Environmental Test Kits
These 'Test N Tube' kits are used for the determination of nitrite by the diazotisation method.
  • Test 'N Tube products provide safe, convenient testing
  • TNT plus vials reduce errors with bar coded vials and RFID
  • TNT plus vials require no reagent blank