Chlorophosphonazo Indicator Solution for Hardness Determination
Supplier: Hach
2589549 2589599
104086-442EA 2195.44 USD
104086-442 104086-444
Chlorophosphonazo Indicator Solution for Hardness Determination
Environmental Test Kits
For the determination of ultra low range hardness by chlorophosphonazo method. Ideal for use with Pour-Thru cell. Requires CDTA reagent.