Reagents for Determination of Silica
Supplier: Hach
199553 2254249 199549 2563000
104085-682EA 106.45 USD
104085-682 104088-192 104088-130 104088-246
Reagents for Determination of Silica
Environmental Test Kits
These reagents used for low and ultra-low range silica determination by the Heteropoly Blue method.