pH Test Kits, Model 17
Supplier: Hach
147006 147008 147009 147011
104085-530PK 200.77 USD
104085-530 104085-532 104085-534 104085-536
pH Test Kits, Model 17
Environmental Test Kits
It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.
Delivery information: Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.