Fluoride ISE Standard Solutions
Supplier: Hach
29149 29153 35949
104086-712EA 65.42 USD
104086-712 104086-714 104086-778
Fluoride ISE Standard Solutions
Standards Anionic Standards Fluoride Standards
  • Traceable to NIST standard reference materials
  • Designed for use with Hach meters portfolio and other meters
  • Reliable packaging
Caution: 1 µg/ml = 1 mg/L