Tubidity Standards for 2100A
Supplier: Hach
2659105 2659110
104086-512EA 778.27 USD
104086-512 104088-266
Tubidity Standards for 2100A
Standards Turbidity Standards
Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results