Color Standards
Supplier: Hach
141410
141453
104085-346PK
196.19
USD
104085-346
104085-348
Color Standards
Standards
Color Reference and Spectroscopy Standards
For accuracy checks in color determinations.
- Simple accuracy checks
- Minimize waste with Voluette ampoules
- Reliable results
The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.