pH Standard Solutions, IUPAC, Certified
Supplier: Hach
S11M001 S11M002 S11M003 S11M005 S11M007 S11M008 S11M004 S11M006
104087-782EA 94.62 USD
104087-782 104087-784 104087-786 104087-788 104087-790 104087-792 104088-648 104088-650
pH Standard Solutions, IUPAC, Certified
Buffers pH Calibration Buffers
Buffers traceable to IUPAC/NIST pH scale.
  • For calibration of pH meters
  • Accuracy at 25 °C: ±0.010 or ±0.05 (pH 12.45)
Each standard is delivered in an airtight package ensuring safe storage. The nominal value of the pH standard is guaranteed until first opening, certified value is guaranteed for two years from the date of issue of the certificate.