Portable Water Sampler, AS950
Supplier: Hach
ASPCXXXC121XX ASPCXXXC161XX
104085-102EA 7933.85 USD
104085-102 104085-104
Portable Water Sampler, AS950
Samplers Water Samplers
These lightweight samplers with low power requirements (12 VDC) have been designed for easy mobility.
  • Desiccant and pump tubing easily accessed
  • Large full-color display for intuitive, single-screen programming, USB upload and download capabilities
  • Copy programs to save time and status screening to eliminate uncertainty
  • Configurable alarms and recorded diagnostics for the measurements (channels): pH, level and power supply
  • Rugged see-through pump cover for rapid inspection
  • FSData Desktop software for data storage
They are suitable for field applications including public environmental or industrial pre-treatment monitoring.
Furthermore, pH/T °C probe or a rain gauge can be connected if you choose the according options.
More models on request.
Certifications: CE and UL. Conforms with legislation on polluting discharges or emissions.
Delivery information: Supplied with bottle set(s) (1 or 24) and strainer with 7.5 m vinyl intake tubing (⅜×⅝ ext.Ø)."