Iris Forceps, 1×2 Teeth
Supplier: WORLD PRECISION INSTRUMENTS LLC
15916-G 15917 15917-G 15916
77661-936EA 64.69 USD
77661-936 77661-938 77661-940 77661-934
Iris Forceps, 1×2 Teeth
Forceps
Iris forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. In the early 1800s, a Johann Friedrich Horner, a German ophthalmologist, began design surgical instruments specifically for use in the eye. The result of his work became one of the earliest iris forceps. Iris forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma.
  • Iris forceps with teeth for grasping fine structures
  • 10 cm (4") Long
  • 0.8 mm Tips
  • 1×2 Teeth
  • Surgical grade stainless steel or premium German steel
One blade has one tooth and the other has two. The blades align so that the teeth fit together. If the tissue to be grasped is slippery or hard to hold, surgical forceps with teeth offer a firmer grasp. However, the teeth may cause some tissue damage. The teeth can puncture or tear delicate tissue, or they could leave indentations or marks on the tissue. For delicate tissues, choose serrated tips.
For hard to hold structures, choose tips with teeth. These Iris Forceps are manufactured with surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.