Microtest Plates, 96-Well
Supplier: SARSTEDT INC
82.1583 82.1583.001 82.1581 82.1581.001 82.1582 82.1582.001
77589-204CS 201.17 USD
77589-204 77589-206 77589-180 77589-182 77589-196 77589-198
Microtest Plates, 96-Well
Microplates HTS Microplates
Microtest plates with flat, round, or conical bases for a variety of applications, such as producing dilution series, quantifying protein concentrations, or storing samples.
  • Made of ultra-pure, crystal-clear polystyrene with three base shapes available
  • Format complies with ANSI/SLAS standard; suitable for all dispensers, washers, and readers
  • Alphanumeric labelling on wells
  • Nonsterile (without lid) and sterile (with lid) formats available
These plates comply with ANSI/SLAS standards and are ideal for conducting parallel analyses on several samples. Alphanumeric labeling, including between wells, makes pipetting easier. Labeling batch number and expiration date on every plate facilitates traceability. All plates are free from DNA/DNase/RNase, free from pyrogens and endotoxins, and non-cytotoxic.