CryoPure Tubes with External Thread
Supplier: SARSTEDT INC
72.379 72.379.002 72.379.004 72.379.005 72.379.006 72.379.007 72.379.992
77588-646CS 914.69 USD
77588-646 77588-648 77588-650 77588-652 77588-654 77588-656 77588-658
CryoPure Tubes with External Thread
Vials
High-quality, safe and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Nominal volume: 1.8 ml
  • Polypropylene tube material
  • HDPE screw caps assembled
  • For long-term storage of cells and cell components in the gas phase of liquid nitrogen
  • QuickSeal mechanism for ergonomic and secure opening and closing with just one rotation
  • Varied color screw caps and color-coded inserts for visual coding and easy identification of samples
  • Use in combination with the CryoRack 40 allows easy handling with one hand
For the storage of cell materials and cell components at temperatures down to –196 °C, SARSTEDT offers the CryoPure certified freezing system to prevent the exposure of samples to additional risks of contamination. Highly transparent CryoPure tubes are available with an external or internal thread, and a range of volumes.
Colored QuickSeal caps and matching inserts provide many color combinations for coding. Assorted color options include a mix of white, red, yellow, green, and purple.