Binocular Loupes
Supplier: WORLD PRECISION INSTRUMENTS LLC
504037 504052 504497 504053 504054 504055 504496
77663-450EA 489.68 USD
77663-450 77663-454 77663-636 77663-456 77663-458 77663-460 77663-634
Binocular Loupes
Magnifiers
Surgical loupes help to alleviate eye strain by enlarging the image when working on tiny subjects or conducting precision operations. They are portable and easier to use than a surgical microscope.
Correct surgical loupes for application involves several factors, including resolution, working distance, field of view, depth of field, magnification, weight and interpupillary distance.
Lightweight loupes are more comfortable for longer periods of use, and they are less likely to slide down your nose as you work. WPI loupes have adjustable interpupillary distance for a correct fit every time.