Powder Pillows, Chromium
Supplier: Hach
1206699
204399
204499
1271099
104085-236PK
125.29
USD
104085-236
104085-712
104085-714
104088-092
Powder Pillows, Chromium
Environmental Test Kits
ChromaVer 3 reagent for hexavalent chromium determination by the Diphenylcarbohydrazide Method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023.