sensION™+ Combination pH Electrodes
Supplier: Hach
LZW5010T97002 LZW5011T97002 LZW5014T97002 LZW5021T97002
104087-680EA 525.4 USD
104087-680 104087-682 104087-684 104087-686
sensION™+ Combination pH Electrodes
Electrodes pH Electrodes
3-in-1 laboratory combination pH electrodes with built-in temperature sensor.
  • 5010T and 5011T are low maintenance electrodes for general applications
  • 5014T and 5021T are high performance electrodes for special or difficult applications
  • For use with Hach sensION+ laboratory pH meters
Delivery information: Supplied with basic user manual. 5014T and 5021T also include 3.0 M KCl electrolyte refill solution (30 ml).