IntelliCAL™ Ion Selective Electrodes, Fluoride
Supplier: Hach
ISEF12101 ISEF12103 ISEF121AP
104087-316EA 2372.91 USD
104087-316 104087-318 104087-320
IntelliCAL™ Ion Selective Electrodes, Fluoride
Electrodes ISE Electrodes
IntelliCAL™ ISEF121 is a digital combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is available with a 1 or 3 meter cable and is intended for laboratory use. Ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications.
Ordering information: Electrode is supplied with integrated temperature sensor, calibration certificate, and basic user manual.
The analysis package contains the IntelliCAL™ ISEF12101 electrode, fluoride ionic strength adjustor powder pillows (100), fluoride 0.5 mg/L standard solution (500 ml), fluoride 1.0 mg/L standard solution (500 ml), and fluoride 2.0 mg/L standard solution (500 ml).