pH Buffer Solutions

Supplier: Hach

1222149 1222220 1222249 1222349
104085-242EA 35.72 USD
104085-242 104085-246 104085-248 104085-252
pH Buffer Solutions
Buffers pH Calibration Buffers

For calibration of pH meters.


Accuracy: pH is ±0.02 at 25 °C


Certifications: Traceable to NIST.


Caution: Please note that the pH 4 and 10 buffers contain formaldehyde <0.1% and the pH 7 buffer contains formaldehyde 0.1 to 1%.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR