Microbiology Test Kit, MEL/MF Portable Laboratory
Supplier: Hach
2569700 2580100
104086-420EA 8839.38 USD
104086-420 104086-426
Microbiology Test Kit, MEL/MF Portable Laboratory
Microbiology Test Systems Microorganism Tests
The Total Coliform MEL Laboratory is designed for analysts who need portable or small-scale microbiology testing capabilities. From a remote field location to a small scale microbiology lab within your existing lab, this MEL offers all the equipment and accessories to perform microbiological analysis.
Ordering information: Required media should be ordered separately.
Delivery information: Kit includes portable incubator and rack, thermometer, Whirlpak bags with thiosulfate (200), filter funnel/pad assemblies (200), alcohol burner (order alcohol separately), vacuum filtration apparatus, colony counter, magnifier and permanent marker.