Amino Acid F for Rapid Liquid Silica Method
Supplier: Hach
2353011 2651155
104086-142KT 75.85 USD
104086-142 104086-502
Amino Acid F for Rapid Liquid Silica Method
Environmental Test Kits
These are used for determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method. Rapid Liquid™ methods offer an economical way to analyse multiple samples and to measure trace concentrations.