Pocket Pro™ Pocket Conductivity/TDS/salt Testers
Supplier: Hach
9531200 9531300 9531400 9531600 9531500
104087-100EA 336.6 USD
104087-100 104087-102 104087-104 104087-106 104088-376
Pocket Pro™ Pocket Conductivity/TDS/salt Testers
Electrochemistry Meters Conductivity Meters
These testers with integrated sensor measure conductivity, TDS, salinity and temperature of general water samples. They are waterproof and they float.
  • Easy calibration steps and built-in diagnostics
  • Easy-to-replace AAA batteries and a convenient sample cup make maintenance and measurements simple
  • Large LCD screen is easy to read
  • Intuitive user interface
  • One year warranty on meter and sensor
Delivery information: All testers are supplied with 4 AAA batteries, lanyard, calibration solution and quick start guide.