Stablcal® Turbidity Standards Calibration Kit for 2100Q
Supplier: Hach
Stablcal® calibration standards kit, for 2100 series portable turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilized formazin turbidity standards are true formazin dilutions developed for use in any turbidimeter.
- Precise, pre-mixed formazin solutions
- Based on formazin primary standards
- Enhanced stability saves time and labour
- Confidence in results