Sulfuric acid 9.6 mol/L (19.2 N)

Supplier: Hach
Danger

203832 203849
104085-702EA 62.82 USD
104085-702 104085-704
Sulfuric acid 9.6 mol/L (19.2 N)
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR