Sulfuric acid 0.01 mol/L (0.02 N) aqueous solution

Supplier: Hach

20342 20353
104085-698EA 34.04 USD
104085-698 104085-700
Sulfuric acid 0.01 mol/L (0.02 N) aqueous solution
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR