SHOWA CS721 Chemical Resistant Gloves
Supplier: Showa
CS721L-09 CS721M-08 CS721S-07 CS721XL-10 CS721XXL-11
76521-176BG 73.87 USD
76521-176 76521-174 76521-172 76521-178 76521-180
SHOWA CS721 Chemical Resistant Gloves
Gloves
The SHOWA CS721 chemical-resistant gloves keep wearers safe from harmful substances, with an elongated forearm for extra protection.
  • Seamless knit, increased dexterity
  • Oil and chemical-resistant
  • Forearm protection
  • Anti-slip grip, robust durability
  • Double coated nitrile
  • Polyester liner, rough grip
A rough finish nitrile palm coating offers enhanced grip, while the powdered liner wicks away perspiration to prevent slipping and discomfort. The durable CS721 also keeps oil and dust out of the glove, ensuring optimum tactile precision.
Trades and Applications: Chemical spray and treatment, light chemical handling in horticulture, agriculture, coating preparation, laboratory, pharma and analysis, painting and spray workshops, fruit and vegetable processing, grain, mill and starch products, washing and cleaning.
Industries: Aerospace, agriculture, chemical, commercial fishing, engineering, food, janitorial, municipal services.
Certifications: EN ISO 374-5:2016, Cat III 0598, EN 388:2016+A1:2018 3132A, EN ISO 374-1:2016+A1:2018/Type JKLMNOPT, EN 407:2004 X1XXXX, Food Contact, EN ISO 21420:2020
Packaging: 12 pair per polybag, 6 polybags per case; 72 pair per case.
Caution: Gloves provide protection from chemical and mechanical hazards shown. Do not use gloves that show signs of wear. If required, cleanse outer surface of glove with running water. Discard used gloves in compliance with local regulations. Do not wear gloves when there is a risk of entanglement by moving parts of machines.