SHOWA CS710 Chemical Resistant Gloves
Supplier: Showa
CS710L-09 CS710M-08 CS710S-07 CS710XL-10 CS710XXL-11
76521-146BG 58.39 USD
76521-146 76521-144 76521-142 76521-148 76521-150
SHOWA CS710 Chemical Resistant Gloves
Gloves
Prioritize comfort and wearer safety with the CS710 chemical resistant gloves. A double-dipped nitrile coating offers robust durability.
  • Chemical and oil resistant
  • Polyester liner
  • Seamless knit
  • Extra grip
  • Natural rubber latex-free
Enhanced grip from the foamed nitrile palm coating provides extra grip, even in oily or slippery environments.
Applications:
- Chemical spray and treatment
- Light chemical handling in horticulture, agriculture
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
- Fruit and vegetable processing
- Grain, mill and starch products
- Washing and cleaning
Certifications: CE cat. III, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type, EN ISO 374-5:2016, EN 407:2004 certified.
Packaging: Pair per polybag: 10, polybags per case: 6, pair per case: 60.