Glass Alcohol Burner, Grobet USA
Supplier: Grobet USA
14.285 14.292
17805-005EA 62.1 USD
17805-005 17805-129
Glass Alcohol Burner, Grobet USA
Heaters Spirit Lamps
Avoid inconvenient work stoppage for refills; the fuel level is always visible through the clear glass base
Molded facets facilitate easy handling. Wick height is adjustable. The threaded metal cap is fitted with a sealing gasket to help prevent leakage. When not in use, the snuff/storage cap reduces fuel evaporation loss. Cotton wick supplied.
Ordering information: Shipped with glass reservoir, wick holder, gasket, wick, and storage cap.