HyFlex® 11-571 Cut-Resistanct Gloves, Latex-Free, Vend Pack
Supplier: Ansell
HyFlex®
11571VP090 11571VP120 11571VP100 11571VP050 11571VP070 11571VP080 11571VP060 11571VP110
76579-500BG 132.42 USD
76579-500 76579-534 76579-490 76579-492 76579-494 76579-550 76579-572 76579-486
HyFlex® 11-571 Cut-Resistanct Gloves, Latex-Free, Vend Pack
Gloves
Ansell's lightest EN ISO Cut D and ANSI/ISEA 105-2024 Cut A4 glove (versus standard EN ISO Cut D and ANSI/ISEA 105-2024 Cut A4-rated gloves) offering 3 times greater cut resistance versus similar gloves made of standard HPPE yarn.
  • Ultra-lightweight design; 20% lighter for unparalleled comfort and dexterity
  • EN ISO Cut D and ANSI/ISEA 105-2024 Cut A4 protection; 3× greater cut resistance
  • Improved FORTIX™ Technology offering up to 20% greater durability
The ultra-lightweight design of HyFlex® 11-571 makes this EN ISO Cut D and ANSI/ISEA 105-2024 Cut A4-rated safety glove 20% lighter, without the use of harsh materials. The gloves provide heightened defense, with 3 times greater cut resistance using Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Offering increased durability for extended use, improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer lasting handling in abrasive conditions. Touchscreen compatibility allows the wearer to work multiple touchscreen devices while keeping gloves on.
Certifications: Ecovadis label, EN 16350:2014, Food Contact, OEKO-TEX® Hohenstein, Dermatologically Tested, ANSI/ISEA 105-2024 ABR Level 4, ANSI/ISEA 105-2024 PUN Level 4, EN 388:2016, EN ISO 21420:2020, ANSI/ISEA 105-2024 Cut Level A4.
Packaging: Gloves are packaged 6 pairs per bag, 144 pairs per carton.