HyFlex® 11-571 Cut-Resistant Gloves, Latex-Free
Supplier: Ansell Healthcare
11571050 11571060 11571070 11571080 11571090 11571100 11571110 11571120
76579-498BG 129.87 USD
76579-498 76579-562 76579-488 76579-554 76579-480 76579-558 76579-506 76579-548
HyFlex® 11-571 Cut-Resistant Gloves, Latex-Free
Gloves
Ansell's lightest EN ISO cut D and ANSI/ISEA 105-2024 cut A4 glove (versus standard EN ISO cut D and ANSI/ISEA 105-2024 cut A4-rated gloves) offering three times greater cut resistance versus similar gloves made of standard HPPE yarn.
  • Ultra-lightweight design
  • 20% lighter for unparalleled comfort and dexterity
  • 3x greater cut resistance
  • Improved FORTIX™ technology offering up to 20% greater durability
The ultra-lightweight design of HyFlex® 11-571 makes this EN ISO cut D and ANSI/ISEA 105-2024 cut A4-rated safety glove 20% lighter, without the use of harsh materials. The gloves provide heightened defense, with three times greater cut resistance using next generation single HPPE yarn developed with INTERCEPT™ cut resistance technology for reliable hand protection. Offering increased durability for extended use, improved FORTIX™ technology delivers up to 20% more abrasion resistance for longer lasting handling in abrasive conditions. Touchscreen compatibility allows the wearer to work multiple touchscreen devices while keeping gloves on.
Certifications: Ecovadis label, EN 16350:2014, food contact, OEKO-TEX® hohenstein, dermatologically tested, ANSI/ISEA 105-2024 ABR level 4, ANSI/ISEA 105-2024 PUN level 4, EN 388:2016, EN ISO 21420:2020, ANSI/ISEA 105-2024 cut level A4.
Packaging: Gloves are packaged 12 pairs per bag, 144 pairs per carton.