Wax Penetration Needle, Humboldt
Supplier: Humboldt
H-1310
52934-163EA 850.53 USD
52934-163
Wax Penetration Needle, Humboldt
Penetrometer Needles
For waxes with 250 or more penetration.
  • Hardened, highly polished stainless steel
  • Has drilled out shaft
Tapered point blunted at tip of truncated cone.
Ø×L: 3.2 x 83 mm (1/8 x 31/4")
Certifications: Meets ASTM specification D1321-61T.