(3-Chloro-2-methoxyphenyl)cyclopropylmethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
416282-1G
416282-5G
416282-500MG
77638-624EA
555
USD
77638-624
77655-632
77650-628
(3-Chloro-2-methoxyphenyl)cyclopropylmethanone ≥95%
(3-Chloro-2-methoxyphenyl)cyclopropylmethanone
Caution: For Research Use Only
Formula:
C₁₁H₁₁ClO₂ Storage Temperature: Ambient |
CAS Number:
2353579-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...