(2-Methyl-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

369075-5G 369075-25G 369075-1G 369075-500MG
77632-968EA 1441 USD
77632-968 77658-976 77642-402 77646-418
(2-Methyl-5-(trifluoromethyl)phenyl)(phenyl)methanone ≥95%
(2-Methyl-5-(trifluoromethyl)phenyl)(phenyl)methanone

Caution: For Research Use Only

Formula: C₁₅H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD22492763
CAS Number: 2733066-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR