Methyl-3-bromo-4-chloro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

35883-5G 35883-1G 35883-25G 35883-500MG
77632-926EA 880 USD
77632-926 77638-332 77658-922 77646-356
Methyl-3-bromo-4-chloro-5-methylbenzoate ≥95%
Methyl 3-bromo-4-chloro-5-methylbenzoate

Caution: For Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD11846081
CAS Number: 1160574-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR