N-2-Propyn-1-ylbenzenesulfonamide ≥95%

Supplier: AOBChem USA

Warning

358215-5G 358215-25G 358215-1G 358215-500MG
77634-922EA 575 USD
77634-922 77658-920 77642-342 77650-350
N-2-Propyn-1-ylbenzenesulfonamide ≥95%
N-2-Propyn-1-ylbenzenesulfonamide

Caution: For Research Use Only

Formula: C₉H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00510807
CAS Number: 13630-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR