2,5-Difluoro-4-isopropoxybenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
33349-5G
33349-1G
33349-25G
33349-500MG
77634-810EA
425
USD
77634-810
77638-150
77658-786
77650-162
2,5-Difluoro-4-isopropoxybenzoic acid ≥95%
2,5-Difluoro-4-isopropoxybenzoic acid
Caution: For Research Use Only
Formula:
C₁₀H₁₀F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD30834053 CAS Number: 2121514-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...