5-Bromo-3-(N-methylaminocarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32900-5G 32900-25G 32900-1G 32900-500MG
77634-798EA 536 USD
77634-798 77658-758 77642-104 77646-106
5-Bromo-3-(N-methylaminocarbonyl)phenylboronic acid ≥97%
5-Bromo-3-(N-methylaminocarbonyl)phenylboronic acid

Caution: For Research Use Only

Formula: C₈H₉BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD30529869
CAS Number: 2121515-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR