Ethyl 4-ethoxy-α-oxo-2-(trifluoromethyl)benzeneacetate ≥95%

Supplier: AOBChem USA

Warning

328493-5G 328493-25G 328493-1G 328493-500MG
77628-628EA 1484.23 USD
77628-628 77635-650 77642-092 77646-104
Ethyl 4-ethoxy-α-oxo-2-(trifluoromethyl)benzeneacetate ≥95%
Ethyl 4-ethoxy-α-oxo-2-(trifluoromethyl)benzeneacetate

Caution: For Research Use Only

Formula: C₁₃H₁₃F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD35179968
CAS Number: 156606-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR