(3-Chloro-2-fluoro-5-methylphenyl)trimethylsilane ≥95%
Supplier: AOBChem USA
|
Warning
|
327319-5G
327319-25G
327319-1G
327319-500MG
77632-752EA
1441
USD
77632-752
77655-076
77642-080
77646-090
(3-Chloro-2-fluoro-5-methylphenyl)trimethylsilane ≥95%
(3-Chloro-2-fluoro-5-methylphenyl)trimethylsilane
Caution: For Research Use Only
Formula:
C₁₀H₁₄ClFSi Storage Temperature: Ambient |
MDL Number:
MFCD35179386 CAS Number: 3002511-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...