2-(3-Fluorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

313188-5G 313188-25G 313188-1G 313188-500MG
77628-540EA 1484.23 USD
77628-540 77658-640 77641-956 77645-966
2-(3-Fluorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(3-Fluorophenyl)-2-methyl-1,3-dioxolane

Caution: For Research Use Only

Formula: 182.1915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30603958
CAS Number: 918525-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR