2-Benzylbenzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
300644-5G
300644-25G
300644-1G
300644-500MG
77634-678EA
591
USD
77634-678
77658-560
77641-842
77649-902
2-Benzylbenzaldehyde ≥95%
2-Benzylbenzaldehyde
Caution: For Research Use Only
Formula:
C₁₄H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD12025180 CAS Number: 32832-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...