1-Iodo-3-(2-methoxyethoxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
296239-5G
296239-25G
296239-1G
296239-500MG
77634-660EA
482
USD
77634-660
77658-536
77641-816
77649-876
1-Iodo-3-(2-methoxyethoxy)benzene ≥97%
1-Iodo-3-(2-methoxyethoxy)benzene
Caution: For Research Use Only
Formula:
C₉H₁₁IO₂ Storage Temperature: Ambient |
MDL Number:
MFCD14647005 CAS Number: 388111-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...