4-Bromo-1-(3-fluorophenyl)-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
278224-5G
278224-1G
278224-25G
278224-500MG
77632-422EA
656
USD
77632-422
77637-708
77654-654
77645-658
4-Bromo-1-(3-fluorophenyl)-1H-pyrazole ≥97%
4-Bromo-1-(3-fluorophenyl)-1H-pyrazole
Caution: For Research Use Only
Formula:
C₉H₆BrFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD21100061 CAS Number: 1353856-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...