2,4-Diiodopyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
27147-5G
27147-1G
27147-25G
27147-500MG
77634-574EA
376
USD
77634-574
77637-644
77658-368
77645-620
2,4-Diiodopyridine ≥95%
2,4-Diiodopyridine
Caution: For Research Use Only
Formula:
C₅H₃I₂N Storage Temperature: Ambient |
MDL Number:
MFCD07437917 CAS Number: 83674-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...