α-Cyclopropyl-2-(phenylmethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259468-5G 259468-1G 259468-500MG
77658-270EA 2764 USD
77658-270 77641-558 77645-516
α-Cyclopropyl-2-(phenylmethoxy)benzenemethanol ≥95%
α-Cyclopropyl-2-(phenylmethoxy)benzenemethanol

Caution: For Research Use Only

Formula: C₁₇H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34875830
CAS Number: 1880814-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR